Please login to your online account to display your discounted pricing

Alfa Aesar™ Molybdenum silicide sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.5% (metals basis)

$603.39 - $603.39

Chemical Identifiers

Chemical Name or Material Molybdenum silicide sputtering target
CAS 12136-78-6
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
Formula Weight 152.13
MDL Number MFCD00151535
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41077KS

 
alfa aesar™
41077KS
1 each Each for $603.39
Description & Specifications

Specifications

Formula Weight 152.13
Chemical Name or Material Molybdenum silicide sputtering target
TSCA TSCA
Density 6.31g/mL
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
Physical Form Target
Assay 99.5%
Assay Percent Notes (metals basis)
MDL Number MFCD00151535
CAS 12136-78-6
Odor Odorless
Color Dark gray
Melting Point 1870° to 2030°C
Quantity 1 each