Learn More
Molybdenum silicide, Mo 61% min, typically 99.5% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01154930
Description
In electrical resistors, in protective coatings at high temperatures, and in engine parts in space vehiclesMolybdenum silicide is used in electric furnaces, glass, steel, electronics and ceramics industries. It finds application in heat treatment materials. It is also used in microelectronics as a contact material. Further, it serves as a dry lubricant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- In electrical resistors, in protective coatings at high temperatures, and in engine parts in space vehicles
Specifications
Molybdenum silicide | |
12136-78-6 | |
250 g | |
MoSi2 | |
molybdenum disilicide, molybdenum silicide, ms silicide, molybdenum-silicide, molybdenum silicide, mo5si3, molybdenum silicide sputtering target, 50.8mm 2.0in dia x 3.18mm 0.125in thick, molybdenum silicide sputtering target, 50.8mm 2.0in dia x 6.35mm 0.250in thick, molybdenum silicide sputtering target, 76.2mm 3.0in dia x 3.18mm 0.125in thick, molybdenum silicide sputtering target, 76.2mm 3.0in dia x 6.35mm 0.250in thick | |
YXTPWUNVHCYOSP-UHFFFAOYSA-N | |
bis($l^{2}-silanylidene)molybdenum | |
6336985 | |
99.5% (typically) | |
Odorless |
1870°C to 2030°C | |
Powder | |
(metals basis) | |
MFCD00151535 | |
Insoluble in acids and aqua regia,however,it is soluble in HF and HNO3Soluble in nitric acid and hydrofluoric acid; Insoluble in water most acids. | |
[Si]=[Mo]=[Si] | |
152.12 | |
152.13 | |
Mo 61% min. | |
6.31 g/mL |
Chemical Identifiers
12136-78-6 | |
152.12 | |
YXTPWUNVHCYOSP-UHFFFAOYSA-N | |
6336985 | |
[Si]=[Mo]=[Si] |
MoSi2 | |
MFCD00151535 | |
molybdenum disilicide, molybdenum silicide, ms silicide, molybdenum-silicide, molybdenum silicide, mo5si3, molybdenum silicide sputtering target, 50.8mm 2.0in dia x 3.18mm 0.125in thick, molybdenum silicide sputtering target, 50.8mm 2.0in dia x 6.35mm 0.250in thick, molybdenum silicide sputtering target, 76.2mm 3.0in dia x 3.18mm 0.125in thick, molybdenum silicide sputtering target, 76.2mm 3.0in dia x 6.35mm 0.250in thick | |
bis($l^{2}-silanylidene)molybdenum |
Safety and Handling
EINECSNumber : 235-231-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only