missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molybdenum Rod, 6.4mm (0.25 in.) dia, 99.95% (Metals basis), Alfa Aesar™

Approx. 15.2cm/50g

Manufacturer:  Thermo Fisher Scientific 1003318

 View more versions of this product

Catalog No. AA1003318



Specifications

Specifications

Molybdenum
7439-98-7
99.95% (Metals basis)
MFCD00003465
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum
95.95
CHEBI:28685
Gray
50g
Dark gray rod
Mo
6+ ion
[Mo]
Odorless
23932
Rod, 6.4mm (0.25 in.) dia.

Chemical Identifiers

7439-98-7
95.95
ZOKXTWBITQBERF-UHFFFAOYSA-N
23932
molybdenum
Mo
MFCD00003465
6+ ion
CHEBI:28685
[Mo]
Documents