missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Molybdenum rod, 5mm (0.2 in.) dia., Puratronic™, 99.97% (metals basis)

Manufacturer:  Alfa Aesar™ 11475FN

 View more versions of this product

Catalog No. AA11475FN



Specifications

Specifications

Molybdenum rod
10 x 150mm
7439-98-7
Mo
6+ ion
[Mo]
95.95
CHEBI:28685
99.97%
≈2g/cm
5mm (0.2 in.) dia.
(metals basis)
MFCD00003465
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum
23932
Rod
Puratronic™

Chemical Identifiers

7439-98-7
95.95
ZOKXTWBITQBERF-UHFFFAOYSA-N
23932
molybdenum
Mo
MFCD00003465
6+ ion
CHEBI:28685
[Mo]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents