missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Molybdenum rod, 3.17mm (0.125 in.) dia., annealed, 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 10032VP

 View more versions of this product

Catalog No. AA10032VP



Specifications

Specifications

Molybdenum rod
5 x 50cm
7439-98-7
Mo
6+ ion
[Mo]
95.95
CHEBI:28685
99.95%
≈0.72g/cm
3.17mm (0.125 in.) dia., Annealed
(metals basis)
MFCD00003465
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum
23932
Rod

Chemical Identifiers

7439-98-7
95.95
ZOKXTWBITQBERF-UHFFFAOYSA-N
23932
molybdenum
Mo
MFCD00003465
6+ ion
CHEBI:28685
[Mo]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents