Please login to your online account to display your discounted pricing

Molybdenum, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical

$85.00 - $113.47

Chemical Identifiers

Chemical Name or Material Molybdenum, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 1313-27-5
CAS 7647-01-0
CAS 7697-37-2
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00003465
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SM113-100

  SDS
fisher scientific company
FLSM113100
Poly Bottle 100mL Each for $85.00
Case of 6 Each for $510.00

SM113-500

  SDS
fisher scientific company
SM113500
Poly Bottle 500mL Each for $113.47
Case of 6 Each for $680.82
Description & Specifications

Specifications

Chemical Name or Material Molybdenum, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 1313-27-5
ChemAlert Storage Symbol White
CAS 7647-01-0
CAS 7697-37-2
DOT Information DOT Class 8, : Corrosive
CAS 7732-18-5
Purity Grade Certified Grade
Physical Form Liquid
Concentration 990 to 1010ppm
MDL Number MFCD00003465
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
Packaging Poly Bottle
Traceability to NIST LOT NUMBER USED MOLYBDENUM REF. STD.
Vapor Pressure 14mmHg at 20°C
Density 1.0g/cm³
Melting Point 0°C
Boiling Point 100°C
Color Colorless
Quantity 100mL
Safety and Handling