missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molybdenum powder, -100 mesh, 99.95% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1003130

 View more versions of this product

Catalog No. AA1003130



Specifications

Specifications

Molybdenum
7439-98-7
99.95% (Metals basis)
MFCD00003465
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum
95.95
CHEBI:28685
Powder
250g
gray powder
Mo
6+ ion
[Mo]
Odorless
23932
95394

Chemical Identifiers

7439-98-7
95.95
ZOKXTWBITQBERF-UHFFFAOYSA-N
23932
molybdenum
Mo
MFCD00003465
6+ ion
CHEBI:28685
[Mo]
Safety and Handling

Safety and Handling

H228: Flammable solid.

EINECSNumber : 231-107-2

Documents