missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molybdenum powder, 99.95% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1003030

 View more versions of this product

Catalog No. AA1003030



Specifications

Specifications

Molybdenum
7439-98-7
10.22g/cm3 at 20°C
Mo
6+ ion
[Mo]
Odorless
23932
Powder
250g
gray powder
99.95% (Metals basis)
MFCD00003465
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum
95.95
CHEBI:28685
Gray

Chemical Identifiers

7439-98-7
95.95
ZOKXTWBITQBERF-UHFFFAOYSA-N
23932
molybdenum
Mo
MFCD00003465
6+ ion
CHEBI:28685
[Mo]
Safety and Handling

Safety and Handling

H228: Flammable solid.

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-107-2

Documents