missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ Molybdenum powder, -22 mesh, Puratronic™, 99.9975% (metals basis excluding W), W <1000ppm
Manufacturer: Alfa Aesar™ 1297288
Specifications
Molybdenum powder | |
7439-98-7 | |
Mo | |
UN3089 | |
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2 | |
[Mo] | |
Odorless | |
23932 | |
Powder | |
Puratronic™ |
2.5g | |
(metals basis excluding W), W <1000ppm | |
MFCD00003465 | |
−22 Mesh | |
ZOKXTWBITQBERF-UHFFFAOYSA-N | |
molybdenum | |
95.95 | |
CHEBI:28685 | |
99.9975% | |
Gray |
Chemical Identifiers
7439-98-7 | |
95.95 | |
ZOKXTWBITQBERF-UHFFFAOYSA-N | |
23932 | |
molybdenum |
Mo | |
MFCD00003465 | |
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2 | |
CHEBI:28685 | |
[Mo] |
Safety and Handling
GHS H Statement
H228
Flammable solid.
GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.
Warning
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
RTECSNumber : QA4680000
TSCA : TSCA