missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Molybdenum powder, -22 mesh, Puratronic™, 99.9975% (metals basis excluding W), W <1000ppm

Manufacturer:  Alfa Aesar™ 1297222

 View more versions of this product

Catalog No. AA1297222



Specifications

Specifications

Molybdenum powder
7439-98-7
Mo
UN3089
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
[Mo]
Odorless
23932
Powder
Puratronic™
100g
(metals basis excluding W), W <1000ppm
MFCD00003465
−22 Mesh
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum
95.95
CHEBI:28685
99.9975%
Gray

Chemical Identifiers

7439-98-7
95.95
ZOKXTWBITQBERF-UHFFFAOYSA-N
23932
molybdenum
Mo
MFCD00003465
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
CHEBI:28685
[Mo]
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.

RTECSNumber : QA4680000

TSCA : TSCA

Documents