missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Molybdenum Pellet, 99.7% (Metals basis), Alfa Aesar™
Approx. 12.5 x 18 x 25mm, HotPressed, density Approx. 8.0g/cm3
Manufacturer: Alfa Aesar 1324222
Specifications
Molybdenum | |
Hot Pressed | |
Dark gray pellet | |
99.7% (Metals basis) | |
MFCD00003465 | |
ZOKXTWBITQBERF-UHFFFAOYSA-N | |
molybdenum | |
95.95 | |
CHEBI:28685 | |
Gray |
100g | |
7439-98-7 | |
10.22g/cm3 at 20°C | |
Mo | |
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2 | |
[Mo] | |
Odorless | |
23932 | |
Pellet |
Chemical Identifiers
7439-98-7 | |
95.95 | |
ZOKXTWBITQBERF-UHFFFAOYSA-N | |
23932 | |
molybdenum |
Mo | |
MFCD00003465 | |
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2 | |
CHEBI:28685 | |
[Mo] |