missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molybdenum Pellet, 99.7% (Metals basis), Alfa Aesar™

Approx. 12.5 x 18 x 25mm, HotPressed, density Approx. 8.0g/cm3

Manufacturer:  Alfa Aesar 1324236

 View more versions of this product

Catalog No. AA1324236



Specifications

Specifications

Molybdenum
Hot Pressed
Dark gray pellet
Mo
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
[Mo]
Odorless
23932
Pellet
500g
7439-98-7
99.7% (Metals basis)
MFCD00003465
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum
95.95
CHEBI:28685
Gray

Chemical Identifiers

7439-98-7
95.95
ZOKXTWBITQBERF-UHFFFAOYSA-N
23932
molybdenum
Mo
MFCD00003465
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
CHEBI:28685
[Mo]
Documents