missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molybdenum(IV) sulfide, 98%, Alfa Aesar™

Hydrogenation and Isomerization Catalyst, Dry Lubricant

Manufacturer:  Alfa Aesar 1221330

 View more versions of this product

Catalog No. AA1221330



Specifications

Specifications

Molybdenum(IV) sulfide
98%
MFCD00003470
molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote z
S=[Mo]=S
160.07
CHEBI:30704
Powder
Black
1317-33-5
MoS2
Black powder
CWQXQMHSOZUFJS-UHFFFAOYSA-N
bis(sulfanylidene)molybdenum
14823
160.08
Odorless
250g

Chemical Identifiers

1317-33-5
160.07
CWQXQMHSOZUFJS-UHFFFAOYSA-N
14823
bis(sulfanylidene)molybdenum
MoS2
MFCD00003470
molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote z
CHEBI:30704
S=[Mo]=S
Safety and Handling

Safety and Handling

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 215-263-9

Documents