missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molybdenum(IV) selenide, 99.9% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1311206

 View more versions of this product

Catalog No. AA1311206



Specifications

Specifications

Molybdenum(IV) selenide
99.9% (metals basis)
MFCD00014216
bis selanylidene molybdenum
MHWZQNGIEIYAQJ-UHFFFAOYSA-N
bis(selanylidene)molybdenum
82894
-325 Mesh Powder
Odorless
>1200°C
12058-18-3
MoSe2
Dark gray powder
Insoluble in water.
[Se]=[Mo]=[Se]
253.892
253.87
6g/cm3
Gray
5g

Chemical Identifiers

12058-18-3
253.892
MHWZQNGIEIYAQJ-UHFFFAOYSA-N
82894
[Se]=[Mo]=[Se]
MoSe2
MFCD00014216
bis selanylidene molybdenum
bis(selanylidene)molybdenum
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c

H301+H331-H373

DOTInformation : Hazard Class: 9, Packing Group: III

EINECSNumber : 235-027-9

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents