Please login to your online account to display your discounted pricing

Molybdenum(IV) selenide, 99.9% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Molybdenum(IV) selenide
CAS 12058-18-3
Physical Form Powder
Assay Percent Range 99.9% (metals basis)
Molecular Formula MoSe2
Formula Weight 253.87
MDL Number MFCD00014216
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1311206

  SDS
alfa aesar
1311206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1311214

  SDS
alfa aesar
1311214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 253.87
Physical Form Powder
Quantity 5g
Color Dark gray
DOT Information Hazard Class: 9, Packing Group: III
EINECS Number 235-027-9
Assay Percent Range 99.9% (metals basis)
Molecular Formula MoSe2
Odor Odorless
Density 6g/cm3
Health Hazard 1 H301-H331-H373-H400-H410: Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
MDL Number MFCD00014216
Chemical Name or Material Molybdenum(IV) selenide
Appearance Dark gray powder
CAS 12058-18-3
Safety and Handling

H301-H331-H373-H400-H410: Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.