missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molybdenum(IV) selenide, 99.9% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1311214

 View more versions of this product

Catalog No. AA1311214



Specifications

Specifications

Molybdenum(IV) selenide
99.9% (metals basis)
MFCD00014216
molybdenum diselenide, molybdenum selenide, molybdenum iv selenide, diselanylidenemolybdenum, bis selanylidene molybdenum, molybdenum diselenide, crystal, molybdenum iv selenide trace metals basis 5g
[Se]=[Mo]=[Se]
253.892
253.87
Odorless
25g
12058-18-3
MoSe2
Dark gray powder
MHWZQNGIEIYAQJ-UHFFFAOYSA-N
bis(selanylidene)molybdenum
82894
Powder
Gray

Chemical Identifiers

12058-18-3
253.892
MHWZQNGIEIYAQJ-UHFFFAOYSA-N
82894
[Se]=[Mo]=[Se]
MoSe2
MFCD00014216
molybdenum diselenide, molybdenum selenide, molybdenum iv selenide, diselanylidenemolybdenum, bis selanylidene molybdenum, molybdenum diselenide, crystal, molybdenum iv selenide trace metals basis 5g
bis(selanylidene)molybdenum
Safety and Handling

Safety and Handling

H301-H331-H373-H400-H410: Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

DOTInformation : Hazard Class: 9, Packing Group: III

EINECSNumber : 235-027-9

Documents