Learn More
Molybdenum(IV) oxide, 99% (metals basis), Thermo Scientific Chemicals
$88.90 - $228.48
Chemical Identifiers
CAS | 18868-43-4 |
---|---|
Molecular Formula | MoO2 |
Molecular Weight (g/mol) | 127.948 |
MDL Number | MFCD00014223 |
InChI Key | QXYJCZRRLLQGCR-UHFFFAOYSA-N |
Synonym | molybdenum dioxide, molybdenum iv oxide, molybdenum oxide, molybdenum iv oxide trace metals basis 25g, mm4 |
PubChem CID | 29320 |
IUPAC Name | dioxomolybdenum |
SMILES | O=[Mo]=O |
Description
Molybdenum(IV) oxide is used to catalyze the dehydrogenation of alcohols. It is also used as an intermediates and processing aids. It finds application in the manufacture of molybdenum nano-wires.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18868-43-4 | |
127.948 | |
QXYJCZRRLLQGCR-UHFFFAOYSA-N | |
29320 | |
O=[Mo]=O |
MoO2 | |
MFCD00014223 | |
molybdenum dioxide, molybdenum iv oxide, molybdenum oxide, molybdenum iv oxide trace metals basis 25g, mm4 | |
dioxomolybdenum |
Specifications
18868-43-4 | |
Powder | |
99% (metals basis) | |
MFCD00014223 | |
QXYJCZRRLLQGCR-UHFFFAOYSA-N | |
dioxomolybdenum | |
29320 | |
99% | |
6.47 g/mL | |
(metals basis) |
Brown to Black | |
25 g | |
MoO2 | |
molybdenum dioxide, molybdenum iv oxide, molybdenum oxide, molybdenum iv oxide trace metals basis 25g, mm4 | |
O=[Mo]=O | |
127.948 | |
127.94 | |
Odorless | |
Molybdenum (IV) oxide |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves. In case of insufficient ventilation, wear suitable respiratory equipment.
H301-H332-H319-H335: Toxic if swallowed. Harmful if inhaled. Causes serious eye irritation. May cause respiratory irritation.
EINECSNumber : 242-637-9
RTECSNumber : QA4688000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only