missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molybdenum(IV) oxide, 99% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04811714

 View more versions of this product

Catalog No. AA4811714



Description

Description

Molybdenum(IV) oxide is used to catalyze the dehydrogenation of alcohols. It is also used as an intermediates and processing aids. It finds application in the manufacture of molybdenum nano-wires.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Molybdenum (IV) oxide
Brown to Black
25 g
(metals basis)
MFCD00014223
QXYJCZRRLLQGCR-UHFFFAOYSA-N
dioxomolybdenum
29320
99%
6.47 g/mL
18868-43-4
Powder
99% (metals basis)
MoO2
molybdenum dioxide, molybdenum iv oxide, molybdenum oxide, molybdenum iv oxide trace metals basis 25g, mm4
O=[Mo]=O
127.948
127.94
Odorless

Chemical Identifiers

18868-43-4
127.948
QXYJCZRRLLQGCR-UHFFFAOYSA-N
29320
O=[Mo]=O
MoO2
MFCD00014223
molybdenum dioxide, molybdenum iv oxide, molybdenum oxide, molybdenum iv oxide trace metals basis 25g, mm4
dioxomolybdenum
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves. In case of insufficient ventilation, wear suitable respiratory equipment.

H301-H332-H319-H335: Toxic if swallowed. Harmful if inhaled. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 242-637-9

RTECSNumber : QA4688000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only