missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Molybdenum(III) chloride, 99.5% (metals basis)

Manufacturer:  Alfa Aesar™ 1403409

 View more versions of this product

Catalog No. AA1403409



Specifications

Specifications

Molybdenum(III) chloride
(metals basis)
MFCD00014214
molybdenum trichloride, molybdenum iii chloride, molybdenum chloride mocl3, molybdenum chloride mocl3, molybdenum iii chloride, mocl3, trichloridomolybdenum, acmc-1bwlx, molybdenum 3+ chloride, mocl3
Cl[Mo](Cl)Cl
202.3
CHEBI:30626
Powder
−100 Mesh
Air & Moisture Sensitive
10g
13478-18-7
Cl3Mo
UN3260
ZSSVQAGPXAAOPV-UHFFFAOYSA-K
trichloromolybdenum
83515
202.32
99.5%
Acidic
Red

Chemical Identifiers

13478-18-7
202.3
ZSSVQAGPXAAOPV-UHFFFAOYSA-K
83515
trichloromolybdenum
Cl3Mo
MFCD00014214
molybdenum trichloride, molybdenum iii chloride, molybdenum chloride mocl3, molybdenum chloride mocl3, molybdenum iii chloride, mocl3, trichloridomolybdenum, acmc-1bwlx, molybdenum 3+ chloride, mocl3
CHEBI:30626
Cl[Mo](Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

RTECSNumber : QA4681400

TSCA : TSCA

Documents