Please login to your online account to display your discounted pricing

Alfa Aesar™ Molybdenum(II) acetate dimer

Chemical Identifiers

Chemical Name or Material Molybdenum(II) acetate dimer
CAS 14221-06-8
Formula Weight 428.06
MDL Number MFCD00013395
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1823906

  SDS
alfa aesar™
1823906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1823903

  SDS
alfa aesar™
1823903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1823914

  SDS
alfa aesar™
1823914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 428.06
Chemical Name or Material Molybdenum(II) acetate dimer
Physical Form Crystalline
MDL Number MFCD00013395
CAS 14221-06-8
Quantity 5g
Sensitivity Air Sensitive