missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Molybdenum(II) acetate dimer

Manufacturer:  Alfa Aesar™ 1823906

 View more versions of this product

Catalog No. AA1823906



Specifications

Specifications

Molybdenum(II) acetate dimer
5g
C8H12Mo2O8
molybdenum ii acetate, tetrakis mu-acetato-o:o' dimolybdenum, molybdenum 2+ tetraacetate, dimolybdenum tetraacetate, tetra aceto dimolybdenum, molybdenum ii acetate dimer,, molybdenum ii acetate dimer 1g, molybdenum ii acetate dimer
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Mo+2].[Mo+2]
428.076
428.06
Air Sensitive
14221-06-8
MFCD00013395
DOOLFANBWPPEGQ-UHFFFAOYSA-J
molybdenum(2+);tetraacetate
84269
Crystalline

Chemical Identifiers

14221-06-8
428.076
DOOLFANBWPPEGQ-UHFFFAOYSA-J
84269
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Mo+2].[Mo+2]
C8H12Mo2O8
MFCD00013395
molybdenum ii acetate, tetrakis mu-acetato-o:o' dimolybdenum, molybdenum 2+ tetraacetate, dimolybdenum tetraacetate, tetra aceto dimolybdenum, molybdenum ii acetate dimer,, molybdenum ii acetate dimer 1g, molybdenum ii acetate dimer
molybdenum(2+);tetraacetate
SDS Documents