Please login to your online account to display your discounted pricing

Molybdenum ICP Standard, 1mL = 1mg Mo (1,000ppm Mo), Ricca Chemical

$123.73 - $123.73

Chemical Identifiers

Chemical Name or Material Molybdenum ICP Standard
CAS 7732-18-5
CAS Min % 95.0
CAS Max % 99.0
CAS 1336-21-6
CAS Min % 2.85
CAS Max % 2.96
CAS 13106-76-8
CAS Min % 0.19
CAS Max % 0.2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PMO1KW-100

 
ricca chemical company
RIPMO1KW100
Poly natural 100mL Each for $123.73

PMO1KW-500

 
ricca chemical company
RIPMO1KW500
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless to slight greenish yellow
Quantity 100mL
Concentration or Composition Notes 1,000ppm Mo
Equivalence 1 mL = 1 mg Mo
CAS 1336-21-6
CAS 13106-76-8
Chemical Name or Material Molybdenum ICP Standard
CAS Min % 95.0
CAS Max % 0.2
CAS Max % 2.96
CAS Min % 2.85
CAS Min % 0.19
CAS Max % 99.0
Packaging Poly natural
CAS 7732-18-5