missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molybdenum ICP Standard, 1mL = 10mg Mo (10,000ppm Mo), Ricca Chemical

Manufacturer:  Ricca Chemical Company RIPMO10KW50

 View more versions of this product

Catalog No. PMO10KW-50

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Molybdenum ICP Standard
94.0,1.93,1.88
Liquid
10,000ppm Mo
Clear colorless to slight greenish yellow
7732-18-5,13106-76-8,1336-21-6
98.0,2.0,1.96
Poly natural
1 mL = 10 mg Mo
50mL

Chemical Identifiers

7732-18-5,13106-76-8,1336-21-6
98.0,2.0,1.96
Molybdenum ICP Standard
94.0,1.93,1.88
Liquid