Please login to your online account to display your discounted pricing

Molybdenum ICP Standard, 1mL = 10mg Mo (10,000ppm Mo), Ricca Chemical


Quantity 100mL
Packaging Poly natural

Manufacturer: ricca chemical company  RIPMO10KW100

 View more versions of this product

Catalog No. PMO10KW-100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless to slight greenish yellow
Quantity 100mL
Chemical Name or Material Molybdenum ICP Standard
CAS 7732-18-5
Equivalence 1 mL = 10 mg Mo
Concentration or Composition Notes 10,000ppm Mo
CAS Min % 1.93
CAS Max % 2.0
CAS Max % 98.0
CAS Max % 1.96
CAS 13106-76-8
CAS Min % 94.0
Packaging Poly natural
CAS Min % 1.88
CAS 1336-21-6

Specifications

Chemical Name or Material Molybdenum ICP Standard
CAS 7732-18-5
CAS Min % 94.0
CAS Max % 98.0
CAS 13106-76-8
CAS Min % 1.93
CAS Max % 2.0
CAS 1336-21-6
CAS Min % 1.88
CAS Max % 1.96
Physical Form Liquid