missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Molybdenum gauze, 40 mesh woven from 0.13mm (0.005 in.) dia. wire

Manufacturer:  Alfa Aesar™ 46606FL

 View more versions of this product

Catalog No. AA46606FL



Specifications

Specifications

Molybdenum gauze
40 mesh woven from 0.13mm (0.005 in.) dia. wire
Mo
40 Mesh
[Mo]
95.95
CHEBI:28685
75 x 75mm
7439-98-7
MFCD00003465
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum
23932
Wire Cloth

Chemical Identifiers

7439-98-7
95.95
ZOKXTWBITQBERF-UHFFFAOYSA-N
CHEBI:28685
[Mo]
Mo
MFCD00003465
23932
molybdenum
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents