missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Molybdenum gauze, 40 mesh woven from 0.13mm (0.005 in.) dia. wire

Manufacturer:  Alfa Aesar™ 46606GT

 View more versions of this product

Catalog No. AA46606GT



Specifications

Specifications

Molybdenum gauze
40 mesh woven from 0.13mm (0.005 in.) dia. wire
Mo
40 Mesh
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum
23932
Wire Cloth
150 x 150mm
7439-98-7
MFCD00003465
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
[Mo]
95.95
CHEBI:28685

Chemical Identifiers

7439-98-7
95.95
ZOKXTWBITQBERF-UHFFFAOYSA-N
23932
molybdenum
Mo
MFCD00003465
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
CHEBI:28685
[Mo]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents