missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Molybdenum foil, 0.5mm (0.02 in.) thick, 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 10045HW

 View more versions of this product

Catalog No. AA10045HW



Specifications

Specifications

Molybdenum foil
150 x 300mm
7439-98-7
Mo
6+ ion, amperit 106.2, atom, mchvl, metco 63, molybdenum, elemental, molybdenum, ion 6+, molybdenum, metallic, tsm1, vi ion
[Mo]
95.95
CHEBI:28685
99.95%
≈51g/100x100mm
0.5mm (0.02 in.) thick
(metals basis)
MFCD00003465
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum
23932
Foil

Chemical Identifiers

7439-98-7
95.95
ZOKXTWBITQBERF-UHFFFAOYSA-N
23932
molybdenum
Mo
MFCD00003465
6+ ion, amperit 106.2, atom, mchvl, metco 63, molybdenum, elemental, molybdenum, ion 6+, molybdenum, metallic, tsm1, vi ion
CHEBI:28685
[Mo]
Safety and Handling

Safety and Handling

EINECSNumber : 231-107-2

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents