missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molybdenum foil, 0.1mm (0.004in) thick, 99.95% (metals basis), Alfa Aesar

Form: approx. 10.2g/100x100mm

Manufacturer:  Thermo Fisher Scientific 10043GJ

 View more versions of this product

Catalog No. AA10043GJ



Specifications

Specifications

Molybdenum
7439-98-7
10.22g/cm3 at 20°C
Mo
6+ ion
[Mo]
Odorless
23932
95394
Gray
100 x 150mm
Dark gray foil
99.95% (Metals basis)
MFCD00003465
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum
95.95
CHEBI:28685
Foil, 0.1mm (0.004 in.) thick

Chemical Identifiers

7439-98-7
95.95
ZOKXTWBITQBERF-UHFFFAOYSA-N
23932
molybdenum
Mo
MFCD00003465
6+ ion
CHEBI:28685
[Mo]
Safety and Handling

Safety and Handling

EINECSNumber : 231-107-2

Documents