missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molybdenum foil, 0.1mm (0.004in) thick, 99.95% (metals basis), Alfa Aesar

Form: approx. 10.2g/100x100mm

Manufacturer:  Alfa Aesar 10043HW

 View more versions of this product

Catalog No. AA10043HW



Specifications

Specifications

Molybdenum
7439-98-7
99.95% (Metals basis)
MFCD00003465
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum
95.95
CHEBI:28685
Foil, 0.1mm (0.004 in.) thick
150 x 300mm
Dark gray foil
Mo
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
[Mo]
Odorless
23932
95394
Gray

Chemical Identifiers

7439-98-7
95.95
ZOKXTWBITQBERF-UHFFFAOYSA-N
23932
molybdenum
Mo
MFCD00003465
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
CHEBI:28685
[Mo]
Safety and Handling

Safety and Handling

EINECSNumber : 231-107-2

Documents