missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molybdenum Foil, 0.025mm (0.001 in.) thick, 99.95% (Metals basis), Alfa Aesar™

Approx. 5.8g/150 x 150mm

Manufacturer:  Thermo Fisher Scientific 10042HZ

 View more versions of this product

Catalog No. AA10042HZ



Specifications

Specifications

Molybdenum
7439-98-7
99.95% (Metals basis)
MFCD00003465
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum
95.95
CHEBI:28685
Gray
300 x 1200mm
Dark gray foil
Mo
6+ ion
[Mo]
Odorless
23932
Foil, 0.025mm (0.001 in.) thick

Chemical Identifiers

7439-98-7
95.95
ZOKXTWBITQBERF-UHFFFAOYSA-N
23932
molybdenum
Mo
MFCD00003465
6+ ion
CHEBI:28685
[Mo]
Documents