missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molybdenum AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Mo), LabChem™

Manufacturer:  LabChem LC174007

 View more versions of this product

Catalog No. LC174007



Specifications

Specifications

Molybdenum AA Standard
12054-85-2,7732-18-5
(NH4)6Mo7O24·4H2O
1235.86
Passes Test
Poly Bottle
Traceable to NIST
Colorless
1000ppm (1mL = 1mg Mo)
99.6,0.4
1000ppm ±10ppm (1mL = 1mg Mo)
Liquid
Certified
Soluble in water
1g/mL
125mL

Chemical Identifiers

12054-85-2
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

Documents
Promotions

Promotions