missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molybdenum AA Standard, 1mL = 1mg Mo (1,000ppm Mo), Ricca Chemical

Manufacturer:  Ricca Chemical Company RIAMO1KW500

 View more versions of this product

Catalog No. AMO1KW-500


 

Specifications Description & Specifications

Specifications

Molybdenum AA Standard
95.0,2.85,0.19
Liquid
1,000ppm Mo
Clear colorless to slight greenish yellow
7732-18-5,1336-21-6,13106-76-8
99.0,2.96,0.2
Poly natural
1 mL = 1 mg Mo
500mL

Chemical Identifiers

7732-18-5,1336-21-6,13106-76-8
99.0,2.96,0.2
Molybdenum AA Standard
95.0,2.85,0.19
Liquid