missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdate Reagent, Taylor Type, 1.2% (w/v) Sodium Molybdate in 4.9% (v/v) Sulfuric Acid, Ricca Chemical
Supplier: Ricca Chemical Company 519025
Specifications
| Molybdate Reagent | |
| 10102-40-6,7732-18-5,7664-93-9 | |
| 1.11,93.0,8.34 | |
| MFCD00149170 | |
| TVXXNOYZHKPKGW-UHFFFAOYSA-N | |
| disodium dioxomolybdenumbis(olate) | |
| 16211258 | |
| Cubitainer | |
| 10 L |
| Taylor Type | |
| 1.07,89.0,8.02 | |
| MoNa2O4 | |
| sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate | |
| [Na+].[Na+].[O-][Mo]([O-])(=O)=O | |
| 205.93 | |
| CHEBI:75213 | |
| 4.9% (v/v) Sulfuric Acid | |
| 1.2% (w/v) Sodium Molybdate |