missing translation for 'onlineSavingsMsg'
Learn More

Molybdate Reagent, Taylor Type, 1.2% (w/v) Sodium Molybdate in 4.9% (v/v) Sulfuric Acid, Ricca Chemical

Supplier:  Ricca Chemical Company RI51901

 View more versions of this product

Catalog No. 5190-1


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Molybdate Reagent
Taylor Type
1.07,89.0,8.02
H4MoNa2O6
sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate
O.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O
241.96
CHEBI:75213
Laboratory
4.9% (v/v) Sulfuric Acid
4 L
1.2% (w/v) Sodium Molybdate
Colorless to Green-Yellow
10102-40-6 , 7732-18-5 , 7664-93-9
1.11,93.0,8.34
MFCD00149170
FDEIWTXVNPKYDL-UHFFFAOYSA-N
disodium dioxomolybdenumbis(olate) dihydrate
16211258
1.2%
Natural Poly Bottle
<1
Liquid

Chemical Identifiers

10102-40-6
241.96
FDEIWTXVNPKYDL-UHFFFAOYSA-N
16211258
disodium dioxomolybdenumbis(olate) dihydrate
H4MoNa2O6
MFCD00149170
sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate
CHEBI:75213
O.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions