Please login to your online account to display your discounted pricing

Alfa Aesar™ Molecular sieves, Y, ammonium ion, powder

Chemical Identifiers

Chemical Name or Material Molecular sieves Y
Name Note Ammonium ion
MDL Number MFCD00131613
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4190122

 
alfa aesar™
4190122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4190114

 
alfa aesar™
4190114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4190136

 
alfa aesar™
4190136
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Molecular sieves Y
TSCA TSCA
Name Note Ammonium ion
Physical Form Powder
MDL Number MFCD00131613
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H335
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P304+P340-P312-P405-P403+P233-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Store in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Hygroscopic
Quantity 100g
Safety and Handling

Warning

GHS P Statement
P261-P304+P340-P312-P405-P403+P233-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Store in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H335
May cause respiratory irritation.