Please login to your online account to display your discounted pricing

Alfa Aesar™ Molecular sieves, titanium oxide based; mesoporous, 25 Angstrom, powder

In oxidation and photocatalysis

Chemical Identifiers

Chemical Name or Material Molecular sieves, titanium oxide based
Name Note Mesoporous
MDL Number MFCD00131613
Synonym Ti-TMS1; Mesoporous titanium oxide based molecular sieve
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4325002

  SDS
alfa aesar™
4325002
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4325004

  SDS
alfa aesar™
4325004
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4325009

  SDS
alfa aesar™
4325009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 3 GHS P Statement
P261-P281-P304+P340-P312-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Molecular sieves, titanium oxide based
TSCA TSCA
Name Note Mesoporous
Physical Form Powder
MDL Number MFCD00131613
Synonym Ti-TMS1; Mesoporous titanium oxide based molecular sieve
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H351-H332
Suspected of causing cancer.
Harmful if inhaled.
Particle Size 25Å
Sensitivity Hygroscopic
Quantity 0.5g
Safety and Handling

Warning

GHS P Statement
P261-P281-P304+P340-P312-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351-H332
Suspected of causing cancer.
Harmful if inhaled.