Please login to your online account to display your discounted pricing

Alfa Aesar™ Molecular sieves, tantalum oxide based; mesoporous, 32 Angstrom, powder

Chemical Identifiers

Chemical Name or Material Molecular sieves, tantalum oxide based
Name Note Mesoporous
MDL Number MFCD00131613
Synonym Ta-TMSI
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42611MD

 
alfa aesar™
42611MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4261106

 
alfa aesar™
4261106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Molecular sieves, tantalum oxide based
TSCA TSCA
Name Note Mesoporous
Physical Form Powder
MDL Number MFCD00131613
Synonym Ta-TMSI
Particle Size 32Å
Quantity 250mg