Please login to your online account to display your discounted pricing

Alfa Aesar™ Molecular sieves, tantalum oxide based; mesoporous, 22 Angstrom, powder

Catalysis, gas adsorption, optoelectronic materials

Chemical Identifiers

Chemical Name or Material Molecular sieves, tantalum oxide based
Name Note Mesoporous
MDL Number MFCD00131613
Synonym Ta TMSI
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4261203

  SDS
alfa aesar™
4261203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4261206

  SDS
alfa aesar™
4261206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA42612MD

  SDS
alfa aesar™
42612MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Molecular sieves, tantalum oxide based
TSCA TSCA
Name Note Mesoporous
Physical Form Powder
MDL Number MFCD00131613
Synonym Ta TMSI
Particle Size 22Å
Sensitivity Hygroscopic
Quantity 1g