Please login to your online account to display your discounted pricing

Alfa Aesar™ Molecular sieves, AW-500, 3-4mm (0.12-0.16 in.) dia.. pellets

$18.08 - $44.45

Chemical Identifiers

Chemical Name or Material Molecular sieves AW-500
Name Note 3 to 4mm (0.12 to 0.16 in.) dia. pellets
MDL Number MFCD00131613
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4702430

  SDS
alfa aesar™
4702430
250g Each for $18.08

AA47024A1

  SDS
alfa aesar™
47024A1
1kg Each for $44.45
Description & Specifications

Specifications

Physical Form Pellet
Chemical Name or Material Molecular sieves AW-500
Name Note 3 to 4mm (0.12 to 0.16 in.) dia. pellets
MDL Number MFCD00131613
Sensitivity Hygroscopic
Color Pale brown
Quantity 250g