Please login to your online account to display your discounted pricing

Molecular sieves 5A, 4 to 8 mesh, ACROS Organics™


Quantity 2.5kg
Packaging Plastic Bottle

Manufacturer: thermo fisher scientific  AC197280025

 View more versions of this product

Catalog No. AC197280025

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Beads
Quantity 2.5kg
Color Beige
Packaging Plastic Bottle
Particle Size 4 to 8 mesh
Water Adsorption 20% min.
Residual Water 1.5% max. (575°C, 3 hrs.)
MDL Number MFCD00131613
CAS 69912-79-4
Chemical Name or Material Molecular sieves 5A
Name Note 5A, 4 to 8 mesh
Bulk Density 660 to 780g/L
Additional Information 2 % max. (550°C)
Health Hazard 1 HYGROSCOPIC

Specifications

Chemical Name or Material Molecular sieves 5A
Name Note 5A, 4 to 8 mesh
CAS 69912-79-4
MDL Number MFCD00131613
Accessories

HYGROSCOPIC

SDS & Safety Documents