missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Molecular sieves 5A, 4 to 8 mesh, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 197285000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agentSpecifications
20% min. | |
500 g | |
Glass bottle | |
1.5% max. (575°C, 3 hrs.) | |
5A, 4 to 8 mesh | |
69912-79-4 | |
MFCD00147627 | |
[Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O | |
202.14 |
Beige | |
4 to 8 mesh | |
Molecular sieves 5A | |
660 to 780g/L | |
Beads | |
AlNaO6Si2 | |
URGAHOPLAPQHLN-UHFFFAOYSA-N | |
aluminum;sodium;dioxido(oxo)silane | |
19758701 |
Chemical Identifiers
69912-79-4 | |
202.14 | |
URGAHOPLAPQHLN-UHFFFAOYSA-N | |
aluminum;sodium;dioxido(oxo)silane |
AlNaO6Si2 | |
MFCD00147627 | |
19758701 | |
[Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O |
Safety and Handling
HYGROSCOPIC
RUO – Research Use Only