missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Molecular sieves, 5A, 3-4mm (0.12-0.16in) dia. pellets, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 033551A1
Description
Molecular sieves, 5A, 3-4mm (0.12-0.16in) dia. Pellets is used as an sorbent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Brown | |
Hygroscopic | |
3 to 4mm (0.12 to 0.16 in.) dia. pellets | |
69912-79-4 | |
MFCD00147627 | |
URGAHOPLAPQHLN-UHFFFAOYSA-N | |
aluminum;sodium;dioxido(oxo)silane | |
19758701 |
1 kg | |
Molecular sieves 5A | |
Pellet | |
AlNaO6Si2 | |
Insoluble in water. | |
[Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O | |
202.14 |
Chemical Identifiers
69912-79-4 | |
202.14 | |
URGAHOPLAPQHLN-UHFFFAOYSA-N | |
aluminum;sodium;dioxido(oxo)silane |
AlNaO6Si2 | |
MFCD00147627 | |
19758701 | |
[Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only