missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Molecular sieves, 5A, 1-2mm (0.04-0.08 in.) dia.. pellets

Manufacturer:  Alfa Aesar™ 87955A1

 View more versions of this product

Catalog No. AA87955A1



Specifications

Specifications

Molecular sieves 5A
Brown
1 to 2mm (0.04 to 0.08 in.) dia. pellets
AlNaO6Si2
URGAHOPLAPQHLN-UHFFFAOYSA-N
aluminum;sodium;dioxido(oxo)silane
19758701
Hygroscopic
1kg
69912-79-4
MFCD00147627
[O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
202.135
Pellet

Chemical Identifiers

69912-79-4
202.135
URGAHOPLAPQHLN-UHFFFAOYSA-N
aluminum;sodium;dioxido(oxo)silane
AlNaO6Si2
MFCD00147627
19758701
[O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
SDS Documents