missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Molecular sieves 4A, pellets, 3.2mm diameter, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 436730050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agentSpecifications
> 20% (55% rH, 20°C) | |
5 kg | |
Molecular sieves 4A | |
650 to 750g/L | |
70955-01-0 | |
MFCD00131613 | |
COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC | |
440.42 |
Beige | |
Plastic drum | |
< 2% (900°C, 2 hrs.) | |
Cylinders | |
C20H25FN2O8 | |
FJUZEZRZKVMAMD-UHFFFAOYNA-N | |
(2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid | |
73906282 |
Chemical Identifiers
70955-01-0 | |
440.42 | |
FJUZEZRZKVMAMD-UHFFFAOYNA-N | |
COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC |
C20H25FN2O8 | |
MFCD00131613 | |
73906282 |
RUO – Research Use Only