missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Molecular sieves, 4A with indicator, -8+12 (ca 2mm) beads

Manufacturer:  Alfa Aesar™ 32256A7

 View more versions of this product

Catalog No. AA32256A7



Specifications

Specifications

Molecular sieves 4A with indicator
Odorless
Blue
−8+12 (≈ 2mm) beads
C20H25FN2O8
UN3077
CC(C(C(=O)OC)C(=O)OC)C(C(=O)O)NC(=O)C(CC1=CC(=CC=C1)F)NC(=O)C
440.424
Beads
−8+12 Mesh
Hygroscopic
5kg
70955-01-0
MFCD00147627
FJUZEZRZKVMAMD-DRZCJDIDSA-N
(2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid
73906282

Chemical Identifiers

70955-01-0
440.424
FJUZEZRZKVMAMD-DRZCJDIDSA-N
(2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid
C20H25FN2O8
MFCD00147627
73906282
CC(C(C(=O)OC)C(=O)OC)C(C(=O)O)NC(=O)C(CC1=CC(=CC=C1)F)NC(=O)C
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H360-H317-H208
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P342+P311-P405-P501a
Wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

SDS Documents