missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Molecular sieves, 4A, 3-5mm (0.12-0.20in) beads, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0546630
Description
Molecular sieves, 4A, 3-5mm (0.12-0.20in) beads are useful in general gas drying applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
250 g | |
Molecular sieves, 4A | |
70955-01-0 | |
MFCD00131613 | |
FJUZEZRZKVMAMD-UHFFFAOYNA-N | |
(2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid | |
73906282 |
Hygroscopic | |
3 to 5mm (0.12 to 0.20 in) beads | |
C20H25FN2O8 | |
Insoluble in cold water,hot water,methanol,diethyl ether,n-octanol and acetone. | |
COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC | |
440.42 |
Chemical Identifiers
70955-01-0 | |
440.42 | |
FJUZEZRZKVMAMD-UHFFFAOYNA-N | |
(2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid |
C20H25FN2O8 | |
MFCD00131613 | |
73906282 | |
COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only