missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Molecular sieves, 4A, 1-2mm (0.04-0.08 in.) dia.. pellets

Manufacturer:  Alfa Aesar™ 87956A1

 View more versions of this product

Catalog No. AA87956A1



Specifications

Specifications

Molecular sieves 4A
Brown
1 to 2mm (0.04 to 0.08 in.) dia. pellets
C20H25FN2O8
FJUZEZRZKVMAMD-DRZCJDIDSA-N
(2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid
73906282
Hygroscopic
1kg
70955-01-0
MFCD00131613
CC(C(C(=O)OC)C(=O)OC)C(C(=O)O)NC(=O)C(CC1=CC(=CC=C1)F)NC(=O)C
440.424
Pellet

Chemical Identifiers

70955-01-0
440.424
FJUZEZRZKVMAMD-DRZCJDIDSA-N
(2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid
C20H25FN2O8
MFCD00131613
73906282
CC(C(C(=O)OC)C(=O)OC)C(C(=O)O)NC(=O)C(CC1=CC(=CC=C1)F)NC(=O)C
SDS Documents