Please login to your online account to display your discounted pricing

Molecular sieves 3A, powder <50 micron, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: fisher scientific acros  214795000

 View more versions of this product

Catalog No. AC214795000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder or Crystals
Quantity 500g
Color Yellow to yellow-green
pH 11.4 (10% max. aq. suspension)
Particle Size 500 Micron Max.
CAS 308080-99-1
Packaging Glass bottle
Chemical Name or Material Molecular sieves 3A
MDL Number MFCD00131613
Residual Water < 1.5% (550°C, 2 hrs.)
Additional Information 3 % max. (550°C, 2 h)
Water Adsorption > 22% (50%rH, 20°C, 24h)
Health Hazard 1 HYGROSCOPIC

Specifications

Chemical Name or Material Molecular sieves 3A
CAS 308080-99-1
MDL Number MFCD00131613
Accessories

HYGROSCOPIC

SDS & Safety Documents