Please login to your online account to display your discounted pricing

Alfa Aesar™ Molecular sieves, 3A, 3-5mm (0.12-0.20in) beads


Quantity 250g

Manufacturer: alfa aesar™  L0535930

 View more versions of this product

Catalog No. AAL0535930

  • / Each
 

Description & Specifications

Chemical Identifiers

Name Note 3 to 5mm (0.12 to 0.20 in) beads
TSCA TSCA
CAS 308080-99-1
Quantity 250g
Chemical Name or Material Molecular sieves, 3A
MDL Number MFCD00131613

Specifications

Chemical Name or Material Molecular sieves, 3A
CAS 308080-99-1
Name Note 3 to 5mm (0.12 to 0.20 in) beads
MDL Number MFCD00131613