Please login to your online account to display your discounted pricing

Molecular Sieve, Activated, Type 5A, 8 to 12 Mesh, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$1,702.70 - $1,702.70

Chemical Identifiers

Chemical Name or Material Molecular Sieve
CAS 1344-01-0
Name Note Activated, Type 5A
Purity Grade BAKER ANALYZED™ Reagent
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003413

 
avantor performance
2709-03
HDPE Pressure Bottom Poly Bottle 250g Case of 12 Each for $1,702.70
Description & Specifications

Specifications

Mesh Size 8 to 12 Mesh
Packaging HDPE Pressure Bottom Poly Bottle
Quantity 250g
Specific Gravity 1L = 2.10kg
Chemical Name or Material Molecular Sieve
Name Note Activated, Type 5A
CAS 1344-01-0
Purity Grade BAKER ANALYZED™ Reagent